Methyl-4-iodo-2-methoxybenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:35387-93-0
:MFCD00052925
Formula:C₉H₉IO₃
:292.07 g/mol
Melting Pt:93…95 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together