ContactUsLinkComponent
Toluene-p-sulphonic anhydride 95%
Corrosive
Toluene-p-sulphonic anhydride 95%
Certificates
undefined
Corrosive
Toluene-p-sulphonic anhydride 95%
Specification Test Results
  • Appearance:
    Off-white to beige to grey Powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Titration by coupling:
    ≥94 %