


2-Hydroxyethyl macrylate 97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:818-61-1
:
Formula:C₅H₈O₃
MDL Number:MFCD00002865
:116.12 g/mol
Boiling Pt:210…215 °C (760 mmHg)
Melting Pt:–60 °C
Density:1.106 g/cm³
Flash Pt:99 °C
Storage Temperature:Ambient
EINECS:212-454-9
UN:2927
ADR:6.1,II
:GHS06
:GHS05
:GHS09
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4470 to 1.4520 (20°C, 589 nm)
- GC:≥96.0 %
- Color scale:≤30 APHA
- Stabilizer:200 to 450 ppm MEHQ
Specification Test Results
Frequently Bought Together