Order Entry
Czech Republic
ContactUsLinkComponent
2-Chloro-4,6-dimethoxy-1,3,5-triazine 98%
Irritant
Corrosive
Environment
2-Chloro-4,6-dimethoxy-1,3,5-triazine 98%
Certificates
undefined
Irritant
Corrosive
Environment
2-Chloro-4,6-dimethoxy-1,3,5-triazine 98%

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    74 to 78 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together