
Ammonium trifluoroacetate 98%
:3336-58-1
:
:C₂H₄F₃NO₂
:MFCD00012615
:131.05 g/mol
:122…126 °C
:222-072-4
:GHS07
:Ambient
- Appearance:White to off-white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:122 to 126 °C
- Titration with HClO4:≥97.5 %
Frequently Bought Together