

2,4-Toluenediisocyanate 75%, Technical Grade
:584-84-9
:26471-62-5
:C₉H₆N₂O₂
:MFCD00002011
:174.16
:115…120 °C (10 mmHg)
:12.5…13.5 °C
:1.22 g/cm³
:127 °C
:Refrigerator
:247-722-4
:2078
:6.1,II
:GHS06
:GHS08
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5660 to 1.5700 (20°C, 589 nm)
- NMR:70 to 90 Tolylene 2,4-diisocyanate
Frequently Bought Together