

p-Tolylhydrazinium chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:637-60-5
:
Formula:C₇H₁₁ClN₂
MDL Number:MFCD00012940
:158.63 g/mol
Melting Pt:241 °C (dec.)
Storage Temperature:Ambient
EINECS:211-295-2
UN:2811
ADR:6.1,III
:GHS07
:GHS08
Specification Test Results
- Appearance:Off-white to beige to light brown Powder or flakes
- Infrared spectrum:Conforms
- Titration with NaOH:≥97.5 %
Specification Test Results
Frequently Bought Together