Ammonium hexafluorophosphate
Catalog # ABCRAB111965.0100
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
About this item
:
:163 g/mol
Merck Index:12,00557
Melting Pt:198 °C
Density:2.18 g/cm³ (18 °C)
EINECS:241-009-1
UN:3260
ADR:8,III
CAS Number:16941-11-0
Formula:H₄F₆NP
MDL Number:MFCD00064642