
tri-Lithium citrate tetrahydrate
Catalog # ABCRAB109113.0500
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
About this item
:
:281.99 g/mol
EINECS:213-045-8
:GHS07
CAS Number:6080-58-6
Formula:C₆H₅Li₃O₇·4H₂O
MDL Number:MFCD00150030