Nalgene® Bottles, Narrow Neck, Round, HDPE, Fluorinated, with Screw Caps
HDPE, fluorinated with PP, fluorinated screw cap.
- Fluorinated bottles and carboys can be used for most acids, alkalis and aggressive organic solvents
- Round shape, non sterile
- Heavy-duty walls are safe, durable and resistant to splitting and punctures
- Fluorocarbon surfaces inside and out
- Leakproof
Fluorinating these HDPE bottles and carboys provides a cost effective alternative to costly fluoropolymer containers.
Manufactured from Nalgene™ for resistance to permeation, paneling, odor emission, flavor and fragrance loss. Provide superior recepticals for solvents, oils, chemicals and agricultural products.
: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP class VI.
- Catalog No:
- NALG312097-0016
- NALG312097-0032
- NALG312097-0004
- NALG312097-0008
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together