
1,4-Dibromo-2,5-difluorobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:327-51-5
:
Formula:C₆H₂Br₂F₂
MDL Number:MFCD00000346
Boiling Pt:96 °C (20 mmHg)
Melting Pt:60…64 °C
:271.89 g/mol
Storage Temperature:Ambient
EINECS:206-317-2
:GHS07
Related Information
Frequently Bought Together