
3-Bromo-p-anisaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34841-06-0
:
Formula:C₈H₇BrO₂
MDL Number:MFCD00016599
:215.05 g/mol
Boiling Pt:108…110 °C (1 mmHg)
Melting Pt:49…53 °C
Storage Temperature:Ambient
EINECS:252-241-8
:GHS07
Related Information
Frequently Bought Together