
9-Methylanthracene
Catalog # EHERC20840000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
Boiling Pt:347 °C (1013 hPa)
Melting Pt:73…76 °C
Density:1.06 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:9,III
:192.26 g/mol
EINECS:212-299-7
UN:3077
:GHS09
CAS Number:779-02-2
Formula:C₁₅H₁₂
MDL Number:MFCD00001261