

9-Methylanthracene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:779-02-2
:
Formula:C₁₅H₁₂
MDL Number:MFCD00001261
Boiling Pt:196…197 °C (12 mmHg)
Melting Pt:76…79 °C
Density:1.066 g/cm³
EINECS:212-299-7
UN:3077
ADR:9,III
:192.26 g/mol
:GHS05
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to orange Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:77 to 80 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together