
9-(Chloromethyl)anthracene 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:24463-19-2
:
Formula:C₁₅H₁₁Cl
MDL Number:MFCD00001263
:226.71 g/mol
Boiling Pt:371 °C (1013 hPa)
Melting Pt:136…141 °C
Storage Temperature:Ambient
EINECS:246-274-7
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Yellow to golden Powder
- Infrared spectrum:Conforms
- Melting point:136 to 141 °C
- HPLC:≥98.0 %
Specification Test Results
Frequently Bought Together