
1-Hexyne for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:693-02-7
:
Formula:C₆H₁₀
MDL Number:MFCD00009504
:82.15 g/mol
Boiling Pt:71 °C (1013 hPa)
Melting Pt:–132 °C
Density:0.7147 g/cm³ (20 °C)
Flash Pt:–20 °C
UN:1993
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:211-736-9
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Solubility:0.36 g/l (25 °C) (experimental)
- Melting Point:-132 °C
- Molar Mass:82.14 g/mol
- Boiling Point:71 - 72 °C (1013 hPa)
- Vapor Pressure:3535 kPa
- Flash Point:-20 °C
- Refractive Index:1.3974 (20 °C, 589 nm)
- Density:0.72 g/cm3 (20 °C)
- Ignition Point:263 °C
Specification Test Results
Frequently Bought Together