Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
 
2,4,4-Trimethyl-2-pentene 98%
 
undefined
 
 
 
 
2,4,4-Trimethyl-2-pentene 98%

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4150 to 1.4170 (20°C, 589 nm)
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together