


3-Chloro-2-(chloromethyl)propene 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1871-57-4
:
Formula:C₄H₆Cl₂
MDL Number:MFCD00000952
:125 g/mol
Boiling Pt:138…140 °C (760 mmHg)
Melting Pt:14 °C
Density:1.08 g/cm³
Flash Pt:36 °C
Storage Temperature:Refrigerator
EINECS:217-489-3
UN:2929
ADR:6.1,II
:GHS06
:GHS02
:GHS09
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4750 to 1.4790 (20°C, 589 nm)
- GC:≥95 %
Specification Test Results
Frequently Bought Together