
trans-2-Hexene 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4050-45-7
:
Formula:C₆H₁₂
MDL Number:MFCD00009473
:84.16 g/mol
Boiling Pt:68 °C (1013 hPa)
Melting Pt:–98.5 °C
Density:0.66 g/cm³
Flash Pt:–20 °C
Storage Temperature:Ambient
EINECS:223-752-3
UN:1993
ADR:3,II
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3920 to 1.3940 (20°C, 589 nm)
- GC:≥98.0 %
Specification Test Results
Frequently Bought Together