ContactUsLinkComponent
Adapters, bent, glass
Adapters, bent, glass
Catalog # LENZ05313044
undefined
Adapters, bent, glass
Catalog # LENZ05313044
Specifications
  • Cone size:
    29/32 NS
  • Socket size:
    1×14/23+1×29/32 RN NS
  • Pk:
    1
Specifications
About this item
DURAN® Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Connector, bent, with cone and lateral socket NS 29/32, with socket NS 14/23 (for thermometer or air leak tube).