Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Tetrachloroethylene ≥98%, TECHNICAL

Tetrachloroethylene ≥98%, TECHNICAL

Health
Environment
Supplier:   VWR Chemicals
Tetrachloroethylene ≥98%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
Tetrachloroethylene ≥98%, TECHNICAL

4 Options Available Below

Product Details & Documents
Synonyms:  Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene
  • CAS Number:127-18-4
  • Formula:C₂Cl₄
  • MDL Number:MFCD00000834
  • Molecular Weight:165.83 g/mol
  • Merck Index:12,09332
  • Boiling Pt:121.1 °C (1013 hPa)
  • Melting Pt:–22 °C
  • Density:1.63 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-825-9
  • UN:1897
  • ADR:6.1,III
Specifications for Product Family

Specification Test Results

  • Assay:
    Min. 98 %
  • Refractive index (20°C):
    1.503 - 1.505
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.