ContactUsLinkComponent
Column head with valve stopcock
  271-3215
undefined
Column head with valve stopcock
  271-3215
  • Cone size:
    29/32 NS
  • Socket size:
    29/32 NS
  • Pk:
    1
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With PTFE valve for easy and accurate reflux adjustment, with thermometer connection NS 14/23 for thermometers with 55 mm built-in length.