ContactUsLinkComponent
Forceps holder
Forceps holder
Catalog # 232-0189
Dodavatel:  RSG
undefined
Forceps holder
Catalog # 232-0189
Dodavatel:  RSG
Specifikace
  • Description:
    Forceps holder
  • W×D×H:
    180×60×135 mm
  • Pk:
    1
Specifikace
O tomto produktu
Made of stainless steel, ESD coated.
  • For safe storage of all forceps used in the lab
  • Ideal for drying forceps after cleaning
  • Acid resistant, anti-static
  • Can be fastened on tables or under cupboards