
Vinyl acetate
Catalog # PROOMM0537.01
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:86.09 g/mol
Merck Index:12,10130
Boiling Pt:72.3 °C (1013 hPa)
Melting Pt:–100 °C
Density:0.932 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:203-545-4
UN:1301
ADR:3,II
:GHS02
CAS Number:108-05-4
Formula:C₄H₆O₂
MDL Number:MFCD00008713