Order Entry
Czech Republic
ContactUsLinkComponent
2-Methylbutane 99+% for spectroscopy
Irritant
Health
Environment
Flammable
2-Methylbutane 99+% for spectroscopy
Certificates
undefined
Irritant
Health
Environment
Flammable
2-Methylbutane 99+% for spectroscopy

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.02 %
  • GC:
    ≥99.0 %
  • UV:
    1 cm cell vs water
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0003 %
  • Acidity:
    ≤0.0005 meq/g

Specification Test Results

Related Information

Frequently Bought Together