Still head for steam distillation
Katalogové číslo LENZ05033044
Dodavatel: Lenz Laborglas GmbH & CO.KG
Specifikace
- Cone size:29/32 NS
- Socket size:29/32 NS
- Pk:1
Specifikace
O tomto produktu
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With two cones NS 29/32 and one DIN thread GL 14.