Safety spectacles for helmets, 3M™ Helmet Integrated Eyewear
Catalog # MMMAV6B
Supplier: 3M
Specifications
- Type:Anti-UV, anti-static, anti-scratch
- Colour:Black
- Description:Integrated Eyewear for Peltor™ helmets
- Seq:01
- Lenses:Grey
- Pk:1
Specifications
About this item
3M™ Integrated Eyewear V6B have a clear lens and are easy to attach to the headband of 3M™ Safety helmets. The unique design allows the glasses to slide easily between the helmet shell and headband.
- Glasses easily attach to the headband of 3M™ Safety helmets for eye protection
- Fit closely to the wearer's cheeks, providing excellent protection from particles and dust
- Durable lens coating helps minimise physical and chemical damage
- Easy to adjust for a good fit
Certifications: EN 166; (EN 172)