4-Hydroxy-4-methyl-2-pentanone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:123-42-2
:
Formula:(CH₃)₂C(OH)CH₂COCH₃
MDL Number:MFCD00004471
:116.16 g/mol
Merck Index:13,02983
Boiling Pt:166 °C (1013 hPa)
Melting Pt:–44 °C
Density:0.94 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-626-7
UN:1148
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:58 °C
REACH:01-2119473975-21
:GHS02
:GHS08
:GHS07
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-47 °C
- Molar Mass:116.16 g/mol
- Boiling Point:168 °C
- Vapor Pressure:1.1 hPa (20 °C)
- Flash Point:58 °C
- Refractive Index:1.4233 (20 °C, 589 nm)
- Explosion Limit:1.8 - 6.9 %(V)
- Density:0.94 g/cm3 (20 °C)
- Ignition Point:640 °C
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Related Information
Frequently Bought Together