Order Entry
Czech Republic
ContactUsLinkComponent
4-Methyl-2-pentanone ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
4-Methyl-2-pentanone ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
4-Methyl-2-pentanone ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Assay:
    Min. 99.0 %
  • IR Spectrum:
    Passes test
  • Acidity:
    Max. 0.002 meq/g
  • Boiling point:
    115 - 117 °C
  • Colouration:
    Max. 10 APHA
  • Density (20/4):
    0.799 - 0.803
  • Density (20/20):
    0.800 - 0.804
  • Non-volatile residue:
    Max. 10 ppm
  • Substances reducing KMnO4 (as O):
    Max. 3 ppm
  • Water:
    Max. 0.1 %
  • Al (Aluminium):
    Max. 0.5 ppm
  • B (Boron):
    Max. 0.02 ppm
  • Ba (Barium):
    Max. 0.1 ppm
  • Ca (Calcium):
    Max. 0.5 ppm
  • Cd (Cadmium):
    Max. 0.05 ppm
  • Co (Cobalt):
    Max. 0.02 ppm
  • Cr (Chromium):
    Max. 0.02 ppm
  • Cu (Copper):
    Max. 0.02 ppm
  • Fe (Iron):
    Max. 0.1 ppm
  • Mg (Magnesium):
    Max. 0.1 ppm
  • Mn (Manganese):
    Max. 0.02 ppm
  • Ni (Nickel):
    Max. 0.02 ppm
  • Pb (Lead):
    Max. 0.1 ppm
  • Sn (Tin):
    Max. 0.1 ppm
  • Zn (Zinc):
    Max. 0.1 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together