

4-Methyl-2-pentanone 98.5+% ACS
:108-10-1
:
:(CH₃)₂CHCH₂COCH₃
:MFCD00008938
:100.16 g/mol
:13,05227
:117.4 °C (1013 hPa)
:–84 °C
:0.8 g/cm³
:14 °C
:Ambient
:203-550-1
:1245
:3,II
:GHS07
:GHS02
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3950 to 1.3970 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥98.5 %
- Color scale:≤15 APHA
- Residue after evaporation:≤0.005 %
- Titrable acid:≤0.002 meq/g
Frequently Bought Together