

2,4-Dimethyl-3-pentanone ≥98%
Catalog # A12875.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:114.19 g/mol
Boiling Pt:124…125 °C
Melting Pt:–69 °C
Density:0.803
Flash Pt:15 °C (59 °F)
Storage Temperature:Ambient
EINECS:209-294-7
UN:1224
ADR:3,II
:GHS02
:GHS07
CAS Number:565-80-0
Formula:C₇H₁₄O
MDL Number:MFCD00008918