Order Entry
Czech Republic
ContactUsLinkComponent
n-Tetratriacontane
n-Tetratriacontane
Catalog # EHERC17430500
Supplier:  EHRENSTORFER
CAS Number:  
n-Tetratriacontane
Catalog # EHERC17430500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    20 mg
  • Seq:
    2305

Specifications

About this item

 :
 :478.93 g/mol
Boiling Pt:482 °C (1013 hPa)
Melting Pt:72.6 °C
Density:0.7728 g/cm³ (90 °C)
Storage Temperature:Ambient
EINECS:238-013-0
CAS Number:14167-59-0
Formula:H₃C(CH₂)₃₂CH₃
MDL Number:MFCD00009412