Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
 
n-Pentane ≥99% (by GC), CHROMASOLV™ AMD, Riedel-de Haën™
 
undefined
 
 
 
 
n-Pentane ≥99% (by GC), CHROMASOLV™ AMD, Riedel-de Haën™

 

  • Refractive Index:
    n20/D 1.358 (lit.)
  • Assay (GC):
    Min. 99 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.005 %
  • Free acid (as CH₃COOH):
    Max. 0.0005 %
  • Transmittance at 200 nm:
    Min. 20 %
  • Transmittance at 210 nm:
    Min. 60 %
  • Transmittance at 220 nm:
    Min. 85 %
  • Transmittance at 230 nm:
    Min. 95 %
  • Transmittance from 250 nm:
    Min. 99 %
  • Fluorescence (chinin) at 254 nm:
    Max. 1 ppb
  • Fluorescence (chinin) at 365 nm:
    Max. 1 ppb
  • for AMD technic:
    complying
  • Vapor Density:
    2.48 (vs air)

 

 

Frequently Bought Together