



n-Pentane, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Dodavatel: Avantor
Certifikáty
O tomto produktu
Číslo CAS:109-66-0
:
Vzorec:H₃C(CH₂)₃CH₃
MDL číslo:MFCD00009498
:72.15 g/mol
Merck Index:13,07193
Bod varu:35…36 °C (1013 hPa)
Bod tání:–130 °C
Hustota:0.63 g/cm³ (20 °C)
Bod vzplanutí:–40 °C
EINECS:203-692-4
UN:1265
ADR:3,II
REACH:01-2119459286-30
:GHS02
:GHS08
:GHS07
:GHS09
Teplota pro skladování:Ambient
Výsledky testů specifikací
- Assay (by GC):min. 99 %
- Boiling Range:35 - 37 °C
- Density (g/ml) at 20°C:0.624 - 0.626
- Residue after Evaporation:max. 0.001 %
- Sulfur Compounds (as S):max. 0.005 %
- Water (H2O):max. 0.01 %
- Trace Impurities (in ppm)Aluminium (Al):max. 0.5
- Trace Impurities (in ppm)Boron (B):max. 0.02
- Trace Impurities (in ppm)Barium (Ba):max. 0.1
- Trace Impurities (in ppm)Calcium (Ca):max. 0.5
- Trace Impurities (in ppm)Cadmium (Cd):max. 0.05
- Trace Impurities (in ppm)Cobalt (Co):max. 0.02
- Trace Impurities (in ppm)Chromium (Cr):max. 0.02
- Trace Impurities (in ppm)Copper (Cu):max. 0.02
- Trace Impurities (in ppm)Iron (Fe):max. 0.1
- Trace Impurities (in ppm)Magnesium (Mg):max. 0.1
- Trace Impurities (in ppm)Manganese (Mn):max. 0.02
- Trace Impurities (in ppm)Nickel (Ni):max. 0.02
- Trace Impurities (in ppm)Lead (Pb):max. 0.1
- Trace Impurities (in ppm)Tin (Sn):max. 0.1
- Trace Impurities (in ppm)Zinc (Zn):max. 0.1
Výsledky testů specifikací
Frequently Bought Together