



n-Pentane 99+% for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:109-66-0
:
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498
:72.15 g/mol
Merck Index:14,7116
Boiling Pt:36 °C
Melting Pt:–130 °C
Density:0.626
Flash Pt:–49 °C (–57 °F)
EINECS:203-692-4
UN:1265
ADR:3,II
REACH:01-2119459286-30
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance (Color):Clear, colorless
- Form:Liquid
- Assay (unspecified) (all isomers):>99.5%
- Assay (unspecified) (n-Pentane):>95%
- Water Content (Karl Fischer Titration):< 50 ppm
Specification Test Results
Related Information
Frequently Bought Together