Order Entry
Czech Republic
ContactUsLinkComponent
cis-1,4-Hexadiene, Sigma-Aldrich®
cis-1,4-Hexadiene, Sigma-Aldrich®
Catalog # SIAL52435-5ML
Supplier:  Merck
CAS Number:  
cis-1,4-Hexadiene, Sigma-Aldrich®
Catalog # SIAL52435-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :82.15 g/mol
EINECS:230-783-6
UN:2458
ADR:3,II
CAS Number:7318-67-4
Formula:C₆H₁₀