2,4-Hexadiene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:592-46-1
:
Formula:C₆H₁₀
MDL Number:MFCD00009297
:82.15 g/mol
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–79 °C
Density:0.7156 g/cm³ (20 °C)
EINECS:209-757-3
ADR:3,II
:M-Clarity™ quality level = MQ100
UN:2458
Frequently Bought Together