4'-Trifluoromethoxyacetophenone, Sigma-Aldrich®
SIAL370592-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:204.15
:110 °C (30 mmHg)
:1.27 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:285-066-0
:85013-98-5
:C₉H₇F₃O₂
:MFCD00042404