Acetophenone-[ring-D5], Sigma-Aldrich®
Catalog # SIAL319856-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:125.11
:M-Clarity™ quality level = MQ200
CAS Number:28077-64-7
Formula:C₈H₃D₅O