trans,trans-2,4-Hexadien-1-ol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:17102-64-6
:
Formula:C₆H₁₀O
MDL Number:MFCD00002925
:98.14 g/mol
:M-Clarity™ quality level = MQ100
EINECS:241-173-4
Frequently Bought Together