
Perfluoroheptane (mixture of isomers) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:335-57-9
:
Formula:C₇F₁₆
MDL Number:MFCD00040339
Boiling Pt:80…82 °C
Density:1.720
:388.05 g/mol
EINECS:206-392-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together