

Cobalt(II) thiocyanate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3017-60-5
:
Formula:C₂CoN₂S₂
MDL Number:MFCD00010937
:175.1 g/mol
Density:1.955 g/cm³ (25 °C)
EINECS:221-156-8
ADR:9,III
:GHS07
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together