

2,4-Hexadiene mixture of isomers 90% tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:592-46-1
:
Formula:C₆H₁₀
MDL Number:MFCD00009297
:82.15 g/mol
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–79 °C
Density:0.72 g/cm³
Flash Pt:–8 °C
EINECS:209-757-3
UN:2458
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥88 %
Specification Test Results
Frequently Bought Together