

trans,trans-2,4-Hexadien-1-ol 99% stabilised
:17102-64-6
:
:C₆H₁₀O
:MFCD00002925
:98.14 g/mol
:80 °C (12 mmHg)
:30.5…31.5 °C
:0.87 g/cm³
:72 °C
:Ambient
:241-173-4
:2811
:6.1,III
:GHS06
:GHS05
- Appearance:Clear colorless to white to light yellow Low melting solid or liquid after melting
- Infrared spectrum:Conforms
- GC:≥98.5 %
- Stabilizer:0.1 % alpha-tocopherol
Frequently Bought Together