Order Entry
Czech Republic
ContactUsLinkComponent
 
Propagyl bromide 80% (w/w) in toluene stabilised
 
undefined
Propagyl bromide 80% (w/w) in toluene stabilised

 

  • Appearance:
    Colorless to yellow-orange Solution
  • Refractive index:
    1.4910 to 1.4970 (20°C, 589 nm)
  • Titration Argentometric:
    75 to 85 % (Product in toluene)
  • Density:
    (20°C) 1.33 to 1.38 g/ml
  • GC:
    ≥96.0 % (Excluding toluene)
  • Stabilizer:
    0.3 % MgO

 

 

Frequently Bought Together