Order Entry
Czech Republic
ContactUsLinkComponent
Filter cartridges, Sartoclean® GF
Filter cartridges, Sartoclean® GF
Catalog # SARP5602504E1
Supplier:  Sartorius
undefined
Filter cartridges, Sartoclean® GF
Catalog # SARP5602504E1
Supplier:  Sartorius

Specifications

  • Filtration area:
    4700 cm²
  • Pore size:
    0,8 μm
  • Type:
    25 (2-Flange bayonet adapter with 226 double O-ring)
  • Seq:
    03
  • Pk:
    1

Specifications

About this item

Sartoclean® filter cartridges are cellulose acetate (CA) membrane based prefilters. These filters are optimised for the protection of downstream equipment and Sartobran® CA final filters.

  • Ultra-low unspecific binding membrane
  • High product yields
  • Increased total throughput performance

The heterogeneous double layer is protected by a layer of glass fleece. This increased the total throughput performance of the membrane double layer and adds an adsorptive component to filtration process.