



n-Hexane ≥99% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:66…69 °C(1.013 hPa)
Melting Pt:–95 °C
Density:0.660…0.668 g/cm³ (20 °C)
Flash Pt:7.4 %(V)
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS07
:GHS08
:GHS02
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Refractive Index:n20/D 1.375 (lit.)
- Assay (GC):Min. 99 %
- Boiling range:68 - 69 °C
- Boiling range 67-69 °C:Min. 95 %
- Density (D 20/4):0.658 - 0.662
- Density (D 20/20):0.659 - 0.663
- Refractive index (n 20/D):1.3745 - 1.3755
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.01 %
- Free acid (as CH₃COOH):Max. 0.002 %
- Aluminium (Al):Max. 0.00005 %
- Boron (B):Max. 0.000002 %
- Barium (Ba):Max. 0.00001 %
- Calcium (Ca):Max. 0.00005 %
- Cadmium (Cd):Max. 0.000005 %
- Cobalt (Co):Max. 0.000002 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000002 %
- Iron (Fe):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000002 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.00001 %
- Tin (Sn):Max. 0.00001 %
- Zinc (Zn):Max. 0.00001 %
- Aromatic substances (as C₆H₆):Max. 0.01 %
- S-compounds (as S):Max. 0.005 %
- Thiophene:Max. 0.0001 %
- Reaction against H₂SO₄:complying
- APHA:Max. 10
Specification Test Results
Frequently Bought Together