ContactUsLinkComponent
n-Hexane, extra dry over Molecular Sieve 97%, AcroSeal®
n-Hexane, extra dry over Molecular Sieve 97%, AcroSeal®
Certificates
undefined
n-Hexane, extra dry over Molecular Sieve 97%, AcroSeal®
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.005 % (Coulometric)
  • GC:
    ≥96.0 %