



n-Hexane 95+%, extra pure
:110-54-3
:92112-69-1
:H₃C(CH₂)₄CH₃
:MFCD00009520
:86.18 g/mol
:13,04712
:69 °C (1013 hPa)
:–95 °C
:0.659 g/cm³
:–22 °C
:203-777-6
:1208
:3,II
:GHS07
:GHS08
:GHS09
:GHS02
:Ambient
- Appearance:Clear liquid
- Water:≤0.05 % (K.F.)
- GC:≥95.0 %
- Color scale:≤20 APHA
- Residue after evaporation:≤0.005 %
Frequently Bought Together