Order Entry
Czech Republic
ContactUsLinkComponent
n-Dodecane ≥99%, GPR RECTAPUR®
Health
n-Dodecane ≥99%, GPR RECTAPUR®
Catalog # 23586.293
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
n-Dodecane ≥99%, GPR RECTAPUR®
Catalog # 23586.293
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Density (20/4):
    0.749 - 0.752
  • Refractive index (20°C):
    1.420 - 1.422

Specification Test Results

About this item

 :
 :170.34 g/mol
Boiling Pt:216 °C (1013 hPa)
Melting Pt:–9,6 °C
Density:0.7487 g/cm³ (20 °C)
Flash Pt:74 °C
Storage Temperature:Ambient
EINECS:203-967-9
 :GHS08
CAS Number:112-40-3
Formula:H₃C(CH₂)₁₀CH₃
MDL Number:MFCD00008969